missing translation for 'onlineSavingsMsg'
Learn More

Human CD28 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18794414
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18794414 25 μg 25µg
18704544 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18794414 Supplier R&D Systems Supplier No. MAB3422SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone 2946A
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P10747
Gene Alias CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD28, Asn19-Pro152, Accession # P10747
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 940
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.