missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human CD276 ELISA Ready-SET-Go!™ Kit
Click to view available options
Quantity:
2 x 96 Tests
Unit Size:
Pack of 2
Description
Includes
- Capture Antibody Concentrate
- Detection Antibody Concentrate
- Human CD276 Standard (lyophilized): 20ng/mL upon reconstitution
- Coating Buffer Concentrate
- Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
- Sample Diluent Y
- pre-titrated Streptavidin-HRP
- Tetramethylbenzidine (TMB) Substrate Solution
Human CD276 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
| Assay Sensitivity | 0.16ng/mL |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 2 x 96 Tests |
| Synonym | B7-H3, B7RP-2, B7RP2, B7H3 |
| Target | CD276 (B7-H3) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction