missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 EUR - 396.00 EUR
Specifications
| Antigen | CD2 |
|---|---|
| Clone | 299813 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724449
![]() |
R&D Systems
MAB1856-SP |
25 μg |
201.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18132571
![]() |
R&D Systems
MAB1856 |
100 μg |
396.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Simple Western.Specifications
| CD2 | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P06729 | |
| 914 | |
| Mouse myeloma cell line NS0-derived recombinant human CD2 Lys25-Asp209 Accession # P06729 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse CD2 or recombinant human CD2F-10 is observed. |
| 299813 | |
| Western Blot 2 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD2, CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| CD2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title