missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CD14 Quantikine QuicKit Control Group 263
Description
- For use as quantitative controls for the determination of human CD14 concentrations in biological fluids.
- Concentrations have been assigned using the R&D Systems™ Human CD14 Quantikine™ QuicKit™ ELISA kit.
- Controls are prepared in a buffered protein base with preservatives.
- They contain recombinant human TNF-α at low and high concentrations.
- Controls are supplied lyophilized.
Specifications
Specifications
| Product Type | ELISA |
| For Use With (Equipment) | Catalog number QK383 |
| Gene ID (Entrez) | 929 |
| Host Species | Human |
| Quantity | 1 Each |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein |
| Target | CD14 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?