missing translation for 'onlineSavingsMsg'
Learn More

Human CCL5/RANTES Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18785909
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18785909 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18785909 Supplier R&D Systems Supplier No. AF278SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 19 publications

CCL5/RANTES Polyclonal specifically detects CCL5/RANTES in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL5/RANTES
Applications Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.4 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13501
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Gene Symbols CCL5
Host Species Goat
Immunogen E. coli-derived recombinant human CCL5/RANTES Ser24-Ser91 Accession # P13501
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6352
Test Specificity Detects human CCL5/RANTES in direct ELISAs. Detects human CCL5/RANTES and mouse CCL5/RANTES Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant canine CCL5/RANTES is observed and approximately 20% cross-reactivity with recombinant cotton rat CCL5/RANTES is observed. In Western blots, no cross-reactivity with recombinant human (rh) CCL3 and rhCCL4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.