missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspase-10 (FLICE 2) Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems AF834
This item is not returnable.
View return policy
Description
Caspase-10/FLICE2 Polyclonal specifically detects Caspase-10/FLICE2 in Human samples. It is validated for Western Blot.
Specifications
| Caspase-10/FLICE2 | |
| Polyclonal | |
| Unconjugated | |
| Apoptotic protease Mch-4, CASP10, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, Caspase10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, Mch4, MCH4ALPS2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 843 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CASP10 | |
| KLH-coupled human Caspase-10 (FLICE 2) synthetic peptide KRTVWGAKQISATSLPTAC | |
| 100 μg | |
| Primary | |
| Detects human and mouse Caspase-10 (FLICE 2) precursor in Western blots. Sensitivity for detection of the small subunit is not known. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction