missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAPS2 (aa 973-1007) Control Fragment Recombinant Protein

Product Code. 30204414
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204414

Brand: Invitrogen™ RP106697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66013 (PA5-66013. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CAPS2 and the related protein CAPS1 encode novel neural/endocrine-specific cytosolic and peripheral membrane proteins. Both are essential components of the synaptic vesicle priming machinery and are required for the Ca2+-regulated exocytosis of secretory vesicles; CAPS-deficient neurons contain no or very few fusion competent synaptic vesicles, causing a selective impairment of fast phasic transmitter release. Like CAPS1, numerous isoforms of CAPS2 are known to exist. These isoforms show distinct expression patterns in the brain. For example, CAPS2b showed high expression in the developing cerebellum. Furthermore, one version of CAPS2 mRNA that lacks exon 3 is detected in some autistic patients, suggesting that the differential expression pattern of CAPS2 is involved in neuronal development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86UW7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 93664
Name Human CAPS2 (aa 973-1007) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A230044C21Rik; Ca++-dependent secretion activator 2; Ca2+ dependent secretion activator 2; Ca2+-dependent activator protein for secretion 2; Cadps2; calcium dependent secretion activator 2; calcium-dependent activator protein for secretion 2; Calcium-dependent secretion activator 2; calcyphosin-2; calcyphosine 2; Calcyphosine-2; calcyphosphine 2; Caps2; CAPS-2; cerebellum postnatal development associated protein 2; Cpd2; KIAA1591; RGD1559440; UG0636c06
Common Name CAPS2
Gene Symbol CADPS2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ALPKVPSLPLNLPQIPNISTASWMPSLYESTNGSA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.