missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Calreticulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
203.00 EUR - 443.00 EUR
Specifications
| Antigen | Calreticulin |
|---|---|
| Clone | 681207 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701817
![]() |
R&D Systems
MAB38982-SP |
25 μg |
203.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743207
![]() |
R&D Systems
MAB38982-100 |
100 μg |
443.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calreticulin Monoclonal specifically detects Calreticulin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Calreticulin | |
| Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| autoantigen Ro, CALR, Calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA, Vasostatin | |
| E. coli-derived recombinant human Calreticulin, Glu18-Leu417, Accession # P27797 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Calreticulin in direct ELISAs. In direct ELISAs, no cross-reactivity with human Vasostatin is observed. |
| 681207 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human | |
| P27797 | |
| 811 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title