missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CACNG6 (aa 66-134) Control Fragment Recombinant Protein

Product Code. 30203123
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203123

Brand: Invitrogen™ RP106718

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66028 (PA5-66028. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Voltage-dependent calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of two known gamma subunit proteins. This particular gamma subunit is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two family members that function as transmembrane AMPA receptor regulatory proteins (TARPs). Alternative splicing results in multiple transcript variants. Variants in this gene have been associated with aspirin-intolerant asthma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXT2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 59285
Name Human CACNG6 (aa 66-134) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310033H20Rik; AW050150; CACNG6; calcium channel, voltage-dependent, gamma subunit 6; calcium voltage-gated channel auxiliary subunit gamma 6; neuronal voltage-gated calcium channel gamma-6 subunit; Voltage-dependent calcium channel gamma-6 subunit
Common Name CACNG6
Gene Symbol CACNG6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NTYKANGSAVCEAAHLGLWKACTKRLWQADVPVDRDTCGPAELPGEANCTYFKFFTTGENARIFQRTTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.