missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C14orf159 (aa 390-481) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65751 (PA5-65751. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
C14orf159 is a protein coding gene.
Specifications
Specifications
| Accession Number | Q7Z3D6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 80017 |
| Name | Human C14orf159 (aa 390-481) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C14orf159; chromosome 14 open reading frame 159; DGLUCY; D-glutamate cyclase, mitochondrial; UNQ2439/PRO5000; UPF0317 protein C14orf159, mitochondrial |
| Common Name | C14orf159 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction