missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C13orf18 Full-length ORF (AAH43488.1, 1 a.a. - 595 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Sequence: MVSNHYFLLCVNLPLREIHTPGPSPSCLGDSLAETTLSEDTTDSVGSASPHGSSEKSSSFSLSSTEVHMVRPGYSHRVSLPTSPRILATSPYPETDSAFFEPSHLTSAADEGAVQVSRRTISSNSFSPEVFVLPVDVEKENAHFYVADMIISAMEKMKCNILSQQQTESWSKEVSGLLGSDQPDSEMTFDTNIKQESGSSTSSYSGYEGCAVLQVSPVTETRTYHDVKEICKCDVDEFVILELGDFNDITETCSCSCSSSKSVTYEPDFNSAELLAKELYRVFQKCWILSVVNSQLAGSLSAAGSIVVNEECVRKDFESSMNVVQEIKFKSRIRGTEDWAPPRFQIIFNIHPPLKRDLVVAAQNFFCAGCGTPVEPKFVKRLRYCEYLGKYFCDCCHSYAESCIPARILMMWDFKKYYVSNFSKQLLDSIWHQPIFNLLSIGQSLYAKAKELDRVKEIQEQLFHIKKLLKTCRFANSALKEFEQVPGHLTDELHLFSLEDLVRIKKGLLAPLLKDILKASLAHVAGCELCQGKGFICEFCQNTTVIFPFQTATCRRCSACRACFHKQCFQSSECPRCARITARRKLLESVASAAT
Specifications
Specifications
| Accession Number | AAH43488.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 80183 |
| Molecular Weight (g/mol) | 92.8kDa |
| Name | C13orf18 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Immunogen | MVSNHYFLLCVNLPLREIHTPGPSPSCLGDSLAETTLSEDTTDSVGSASPHGSSEKSSSFSLSSTEVHMVRPGYSHRVSLPTSPRILATSPYPETDSAFFEPSHLTSAADEGAVQVSRRTISSNSFSPEVFVLPVDVEKENAHFYVADMIISAMEKMKCNILSQQQTESWSKEVSGLLGSDQPDSEMTFDTNIKQESGSSTSSYSGYEGCAVLQVSPVTETRTYHDVKEICKCDVDEFVILELGDFNDITETCSCSCSSSKSVTYEPDFNSAELLAKELYRVFQKCWILSVVNSQLAGSLSAAGSIVVNEECVRKDFESSMNVVQEIKFKSRIRGTEDWAPPRFQIIFNIHPPLKRDLVVAAQNFFCAGCGTPVEPKFVKRLRYCEYLGKYFCDCCHSYAESCIPARILMMWDFKKYYVSNFSKQLLDSIWHQPIFNLLSIGQSLYAKAKELDRVKEIQEQLFHIKKLLKTCRFANSALKEFEQVPGHLTDELHLFSLEDLVRIKKGLLAPLLKDILKASLAHVAGCELCQGKGFICEFCQNTTVIFPFQTATCRRCSACRACFHKQCFQSSECPRCARITARRKLLESVASAAT |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction