missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human beta-Glucuronidase/GUSB ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human beta-Glucuronidase/GUSB quantitates human beta-Glucuronidase/GUSB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human beta-Glucuronidase/GUSB.
The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).
Specifications
Specifications
| Accession Number | Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; GUSB; Gus-r; Gus-s; Gus-t; Gus-u; Gut; MPS7 |
| Assay Range | 0.18 to 40 ng/mL |
| Assay Sensitivity | 0.18 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2990 |
| Gene Symbol | GUSB |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction