missing translation for 'onlineSavingsMsg'
Learn More

Human Bcl-2 Minus C-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18733318
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18733318 25 μg 25µg
18112871 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18733318 Supplier R&D Systems Supplier No. MAB8271SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl-2 Monoclonal specifically detects Bcl-2 in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Neutralization
Classification Monoclonal
Clone 118712
Conjugate Unconjugated
Dilution Neutralization
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl2, Bcl-2
Gene Symbols BCL2
Host Species Mouse
Immunogen E. coli-derived recombinant human Bcl-2 minus the carboxyl-terminal mitochondrial targeting sequence
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 596
Test Specificity Detects human Bcl-2 Minus C-Terminus. Neutralizes recombinant human (rh) Bcl-2 bioactivity but not rhBcl-w or rhBcl-x bioactivity.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.