missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 EUR - 467.00 EUR
Specifications
| Antigen | B7-H6 |
|---|---|
| Clone | 1018457 |
| Dilution | Blockade of Receptor-ligand Interaction |
| Applications | Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730617
![]() |
R&D Systems
MAB71444-SP |
25 μg |
188.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18710887
![]() |
R&D Systems
MAB71444-100 |
100 μg |
467.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Blocking AssaySpecifications
| B7-H6 | |
| Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1 | |
| Human embryonic kidney cell HEK293-derived human B7-H6, Asp25-Ser262, Accession # Q68D85 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1018457 | |
| Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q68D85 | |
| 374383 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title