missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human APOBEC3H (aa 85-183) Control Fragment Recombinant Protein

Product Code. 30205719
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205719

Brand: Invitrogen™ RP92353

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54426 (PA5-54426. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DNA deaminase (cytidine deaminase) which acts as an inhibitor of retrovirus replication and retrotransposon mobility via deaminase-dependent and -independent mechanisms. The A3H-var/haplotype 2 exhibits antiviral activity against vif-deficient HIV-1. After the penetration of retroviral nucleocapsids into target cells of infection and the initiation of reverse transcription, it can induce the conversion of cytosine to uracil in the minus-sense single-strand viral DNA, leading to G-to-A hypermutations in the subsequent plus-strand viral DNA. The resultant detrimental levels of mutations in the proviral genome, along with a deamination-independent mechanism that works prior to the proviral integration, together exert efficient antiretroviral effects in infected target cells. Selectively targets single-stranded DNA and does not deaminate double-stranded DNA or single- or double-stranded RNA. Exhibits antiviral activity also against T-cell leukemia virus type 1 (HTLV-1) and may inhibit the mobility of LTR and non-LTR retrotransposons.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6NTF7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 164668
Name Human APOBEC3H (aa 85-183) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A3H; APOBEC3H; APOBEC-related protein 10; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3 H; apolipoprotein B mRNA editing enzyme catalytic subunit 3 H; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3 H; Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3 H; ARP10; ARP-10; DNA dC->dU-editing enzyme APOBEC-3 H
Common Name APOBEC3H
Gene Symbol APOBEC3H
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CSSCAWELVDFIKAHDHLNLGIFASRLYYHWCKPQQKGLRLLCGSQVPVEVMGFPEFADCWENFVDHEKPLSFNPYKMLEELDKNSRAIKRRLERIKQS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.