Learn More
Human alpha-Methylacyl-CoA Racemase/AMACR Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF7508-SP
Product Code. 18758087
  • 201.00 EUR / 25µg
Description
alpha-Methylacyl-CoA Racemase/AMACR Polyclonal specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Methylacyl-CoA Racemase/AMACR
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, alphaMethylacylCoARacemase, AMACR, AMACRD, CBAS4, EC 5.1.99.4, RACE, RM
Sheep
Affinity Purified
RUO
23600
Sterile PBS to a final concentration of 0.2 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.2 ug/mL
Q9UHK6
AMACR
E. coli-derived recombinant human alpha-Methylacyl-CoA Racemase/AMACR Met1-Leu382 Accession # Q9UHK6
25 μg
Primary
Detects human alpha-Methylacyl-CoA Racemase/AMACR in direct ELISAs and Western blots.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions