Learn More
Human alpha-Methylacyl-CoA Racemase/AMACR Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7508-SP
This item is not returnable.
View return policy
Description
alpha-Methylacyl-CoA Racemase/AMACR Polyclonal specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| alpha-Methylacyl-CoA Racemase/AMACR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, alphaMethylacylCoARacemase, AMACR, AMACRD, CBAS4, EC 5.1.99.4, RACE, RM | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23600 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| Q9UHK6 | |
| AMACR | |
| E. coli-derived recombinant human alpha-Methylacyl-CoA Racemase/AMACR Met1-Leu382 Accession # Q9UHK6 | |
| 25 μg | |
| Primary | |
| Detects human alpha-Methylacyl-CoA Racemase/AMACR in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction