missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AICL/CLEC-2B Antibody, R&D Systems™

Mouse Monoclonal Antibody
252.00 EUR - 570.00 EUR
Specifications
| Antigen | AICL/CLEC-2B |
|---|---|
| Clone | 1064935 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18183081
|
R&D Systems
MAB9059-100 |
100 μg |
570.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18152521
![]() |
R&D Systems
MAB9059-SP |
25 μg |
252.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AICL/CLEC-2B Monoclonal antibody specifically detects AICL/CLEC-2B in Human samples. It is validated for Flow CytometrySpecifications
| AICL/CLEC-2B | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Activation-induced C-type lectin, AICLC-type lectin superfamily member 2, CLEC2B, CLEC-2B, CLECSF2, CLECSF2IFN-alpha-2b-inducing-related protein 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 2 (activation-induced), C-type lectin domain family 2 member B, C-type lectin domain family 2, member B, HP10085, IFN-alpha2b-inducing related protein 1, IFNRG1 | |
| Chinese Hamster Ovary cell line, CHO-derived human AICL/CLEC-2B, Lys26-His149, Accession # Q92478 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1064935 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q92478 | |
| 9976 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title