missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adrenomedullin/ADM Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 EUR - 457.00 EUR
Specifications
| Antigen | Adrenomedullin/ADM |
|---|---|
| Clone | 1003019 |
| Dilution | Immunohistochemistry 5-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18744667
![]() |
R&D Systems
MAB61081-SP |
25 μg |
201.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700907
![]() |
R&D Systems
MAB61081-100 |
100 μg |
457.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adrenomedullin/ADM Monoclonal specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry.Specifications
| Adrenomedullin/ADM | |
| Immunohistochemistry 5-25 μg/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADM, adrenomedullin, AMpreproadrenomedullin | |
| Human Adrenomedullin/ADM synthetic peptide, Accession # P35318 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Adrenomedullin/ADM in direct ELISAs. |
| 1003019 | |
| Immunohistochemistry | |
| Unconjugated | |
| Human | |
| P35318 | |
| 133 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title