missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adiponectin/Acrp30 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB10652-SP
This item is not returnable.
View return policy
Description
Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot.
Specifications
| Adiponectin/Acrp30 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q15848 | |
| ADIPOQ | |
| Mouse myeloma cell line NS0-derived recombinant human Adiponectin/Acrp30 Glu19-Asn244 Accession # Q15848 | |
| 25 μg | |
| Primary | |
| Detects human Adiponectin/Acrp30 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Acrp30 (aa 101-244), recombinant mouse (rm) Acrp30 (aa 111-247), rmAcrp30 (HWM), recombinant rat Acrp30 (aa 1-244), rhC1qTNF1, rmCORS26/C1qTNF3, rhC1qTNF4, rhC1qTNF5, rhC1qTNF9, or rhC1qL2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 553517 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9370 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction