missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM23 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4974-SP
This item is not returnable.
View return policy
Description
ADAM23 Monoclonal specifically detects ADAM23 in Human samples. It is validated for Immunohistochemistry.
Specifications
| ADAM23 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O75077 | |
| ADAM23 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM23 Ser60-His585 Accession # O75077 | |
| 25 μg | |
| Primary | |
| Detects human ADAM23 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 504425 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloproteinase domain 23, ADAM metallopeptidase domain 23, disintegrin and metalloproteinase domain-containing protein 23, MDC3, MDC-3, MDC3ADAM 23, MDC-L, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8745 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction