missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Activin R2B/ACVR2B ELISA Kit

Product Code. 17144843
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17144843 96 Tests Pack of 96
17105329 5 x 96 Tests Each
18557954 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17144843 Supplier Invitrogen™ Supplier No. EH8RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Activin R2B/ACVR2B quantitates human Activin R2B/ACVR2B in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Activin R2B/ACVR2B.

ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases and is involved in the activin and myostatin signaling pathway. Activins and inhibins are members of the TGF-beta superfamily due to amino acid homology with respect to the conservation of 7 of the 9 cysteine residues common to all TGF-beta forms. To date, seven type I and five type II activin receptors have been cloned from mammals, including activin receptor IA, activin receptor IIA, activin receptor IB, and activin receptor IIB. In addition, two splice variants of activin receptor IIA and five splice variants of activin receptor IIB have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number activin A receptor type 2 B; activin A receptor type IIB; activin A receptor, type IIB; activin receptor IIB; activin receptor type IIB; Activin receptor type-2 B; Activine receptor 2 b (transmembrane serine kinase); ACTRIIB; ACTR-IIB; ActRIIB type II activin receptor B; ACVR2B; HT x 4; MGC116908
Assay Range 1.3 to 300 ng/mL
Assay Sensitivity 1.3 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 93
Gene Symbol ACVR2B
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target ACVR2B
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.