Learn More
HTR1B (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111905
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: CSAKDYIYQD SISLPWK Homology: Rat - 15/17 amino acid residues identical; mouse - 14/17 amino acid residues identical.
Serotonin (5-hydroxytryptamine, 5-HT), originally discovered as a serum factor plays important roles in regulating diverse biological processes in central and peripheral nervous systems, cardiovascular systems, and gastrointestinal systems. Molecular cloning studies have identified 5 receptors belonging to the 5-HT1 subtypes. These receptors are negatively coupled to adenylyl cyclase. Radioligand binding studies suggest the presence of 5-HT1B (formerly known as 5-HT1DbR) sites in substantia nigra, dorsal subiculum, globus pallidus, and superior collicum. In situ hybridization studies show abundance of 5-HT1BR mRNA in CA1 pyramidal cells of hippocampus, Purkinje cells of cerebellum, olfactory tubercle, subthalamic nucleus and somewhat lower levels in the caudate-putamen, cortical areas, thalamic and hypothalamic nuclei.
Specifications
| HTR1B (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| HTR1B | |
| 5ht1b; 5-HT1B; 5-HT-1B; 5HT1B Receptor; 5-HT1DB; 5-HT-1D-beta; 5-hydroxytryptamine receptor 1B; HTR1B; HTR1D2; HTR1DB; S12; Serotonin 1D beta receptor; Serotonin receptor 1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15551, 25075, 3351 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P28222, P28334, P28564 | |
| HTR1B | |
| Peptide corresponding to amino acid residues 33-49 of human 5-Hydroxytryptamine receptor 1B (Extracellular, N-terminus) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.