Learn More
Tocris Bioscience™ HTH 01-015
GREENER_CHOICE
Potent and selective NUAK1 inhibitor
Brand:  Tocris Bioscience™ 5622/50
Encompass
Product Code. 15776986
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
HTH 01-015 is a potent and selective NUAK1 inhibitor (IC50 = 100 nM). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells in vitro, to a similar extent as NUAK1 knockdown models.
Specifications
Specifications
5,13-Dihydro-4,5,13-trimethyl-2-[[1-(4-piperidinyl)-1H-pyrazol-4-yl]amino]-6H-naphtho[2,3-e]pyrimido[5,4-b][1,4]diazepin-6-one
1613724-42-7
50 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions