Learn More
Invitrogen™ HSC70 Monoclonal Antibody (BB70)

Description
MA1-10891 detects Heat Shock Protein 70/Heat Shock Cognate Protein 70 in human, mouse, rat, rabbit, monkey, hamster, guinea pig, dog, bovine, sheep, pig, chicken, yeast, fish and Xenopus samples. MA1-10891 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, MA1-10891 is specific for the ~72 kDa Heat Shock Protein 70/Heat Shock Cognate Protein 70 protein. MA1-10891 immunogen is chicken Hsp70/Hsp90 complex.
Specifications
Specifications
| Antigen | HSC70 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | BB70 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | Hspa8 |
| Gene Accession No. | O73885, P11142, P63017, P63018 |
| Gene Alias | 2410008N15Rik; AAG2; constitutive heat shock protein 70; constitutively expressed heat shock protein; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; FAM10A1; Heat shock 70 kDa protein 8; heat shock 70kd protein 10; heat shock 70kd protein 10 (HSC71); heat shock 70kD protein 8; heat shock 70kDa protein 8; heat shock cognate; heat shock cognate 70; heat shock cognate 71 kD protein; heat shock cognate 71 kDa protein; heat shock cognate 71 kDa protein-like protein; heat shock cognate 71-kd protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 70 cognate; heat shock protein 8; heat shock protein A8; heat shock protein cognate 70; heat shock protein family A (Hsp70) member 8; HEL-33; HEL-S-72p; HIP; HSC54; Hsc70; Hsc70 ATPase; HSC70 protein; Hsc71; Hsc73; HSP70; HSP71; HSP73; Hspa10; Hspa8; hspa8 protein; I79_022974; LAP; LAP1; LAP-1; lipopolysaccharide-associated protein 1; LPS-associated protein 1; MGC131511; MGC29929; NIP71; N-myristoyltransferase inhibitor protein 71; Protein FAM10A1; similar to 7O KDa heat shock cognate protein; SNC6; unnamed protein product; wu:fb01g06; wu:fi48b06 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.