Learn More
HPRT1 Mouse anti-Human, Unconjugated, Clone: 1C3D1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67518-1-IG-150UL
 View more versions of this product
Product Code. 16810196
  • 410.00 EUR / 150µL
Description
The HPRT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HPRT1. This antibody recognizes human, mouse, rat antigen. The HPRT1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
HPRT1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, ELISA
1C3D1
Unconjugated
150 μL
3251
IgG1