Learn More
HPRT1 Mouse anti-Human, Unconjugated, Clone: 1C3D1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67518-1-IG-150UL
Description
The HPRT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HPRT1. This antibody recognizes human, mouse, rat antigen. The HPRT1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| HPRT1 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 1C3D1 | |
| Unconjugated | |
| 150 μL | |
| 3251 | |
| IgG1 |