Learn More
SCAT™ HPLC Air Valve, for PFAS Analytic
Shop All SCAT Europe Products
Click to view available options
Quantity:
1 Ea.
10 Pk.
8 Pk.
Unit Size:
1 sheet
10 sets
8 sets
Description
- With Luer-Lock connection for gassing the solvents and change indicator for timely change.
- Made of PP for PFAS analysis of polyfluorinated chemicals (formerly: PFC)
- UNF 1/4 in. 28G thread size fits to any SCA PFAS-Analytic Safety Cap for PFAS Analysis
Specifications
Specifications
| For Use With (Equipment) | SCAT Safety Caps for PFAS-Analytic |
| Material | PP |
| Thread Size | UNF 1/4 in. 28G |
| For Use With (Application) | HPLC analysis |
| Type | Valve |
| Quantity | 8 Pk. |
| Color | Red, Green |