missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HP1 alpha Recombinant Rabbit Monoclonal Antibody (8C3M6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535397
This item is not returnable.
View return policy
Description
Immunogen sequence: SNFSNSADDI KSKKKREQSN DIARGFERGL EPEKIIGATD SCGDLMFLMK WKDTDEADLV LAKEANVKCP QIVIAFYEER LTWHAYPEDA ENKEKETAKS.
Heterochromatin protein-1 (HP1) is a methyl-lysine binding protein localized at heterochromatin sites, where it mediates gene silencing. It has been shown that mammalian methyltransferases that selectively methylate histone H3 on lysine-9 generate a binding site for HP1 proteins, a family of heterochromatic adaptor molecules implicated in both gene silencing and supranucleosomal chromatin structure. High-affinity in vitro recognition of a methylated histone H3 peptide by HP1 requires a functional chromodomain. Thus, the HP1 chromodomain is a specific interaction motif for the methyl epitope on lysine-9 of histone H3. In vivo, heterochromatin association of HP1 proteins is lost in Suv39h double-null primary mouse fibroblasts but is restored after reintroduction of a catalytically active SUV39H1 HMTase.
Specifications
| HP1 alpha | |
| Recombinant Monoclonal | |
| 0.64 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P45973, Q61686 | |
| Cbx5 | |
| A synthetic peptide corresponding to a sequence within amino acids 92-191 of human HP1 alpha/CBX5 (P45973). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 8C3M6 | |
| Unconjugated | |
| Cbx5 | |
| 2610029O15Rik; antigen p25; C75991; Cbx5; Cbx5 antibody; chromobox 5; chromobox homolog 5; chromobox homolog 5 (HP1 alpha homolog, Drosophila); chromobox protein homolog 5; chromobox-like protein 5; epididymis luminal protein 25; HEL25; heterochromatin protein 1 alpha; heterochromatin protein 1 homolog alpha; heterochromatin protein 1-alpha; HGNC:1555; HP1; HP1 alpha; HP-1 alpha antibody; HP1 alpha homolog; Hp1a; Hp1alpha; HP1-ALPHA; HP1Hs alpha; HP1Hs-alpha; mHP1(alpha) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12419, 23468, 300266 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction