Learn More
Honeywell™ Howard Leight FirmFit Uncorded Disposable Foam Earplugs
Description
- Longer wear time comfort: More protection. Greater compliance. Greater Protection
- SNR 37dB provides excellent protection in most medium-to-high noise environments
- Easy to Insert: Requires less pressure to roll down, and slow recovery rate gives you plenty of time to correctly insert in ear canal.
- Easy to See: Distinct warning orange color makes compliance checks easy.
Specifications
Specifications
| Color | Orange |
| Certifications/Compliance (Europe) | EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: CEC40030, EC Attestation |
| Packaging Type | Poly bag |
| Pairs per Package | 1 Pair/Polybag |
| Quantity | 200 Pairs/Box, 10 Boxes/Cs. |
| Noise Reduction Rating (NRR) | SNR: 37 dB |
| Plug Shape | Cylinder |
| Disposable | Single-use |
| Material | PVC Foam |