Learn More
Benchmark Scientific™ Horizontal Tube Holder for Benchmark RotoBot™ Series
Product Code. 17984422
Click to view available options
Tube Capacity:
6 x 15 mL Horizontal
6 x 50 mL Horizontal
Unit Size:
Each
Product Code. 17984422
Brand: Benchmark Scientific™ R4040HZ50
Only null left
Add to basket
Add to basket
This tube holder is for the Benchmark RotoBot™ Series and holds the tubes horizontally.
  • Horizontal orientation for rolling type mixing
  • For use with RotoBot™ Series Rotators
  • Specifications
    Tube Capacity 6 x 50 mL Horizontal
    For Use With (Equipment) RotoBot™ Series