Learn More
Scientific Industries SI™ Horizontal Tube Holder
Snap tubes into place swiftly and easily. Scientific Instruments SI™ Horizontal Tube Holder does not require additional attachments for use.
105.00 EUR - 207.06 EUR
Specifications
For Use With (Equipment) | Vortex-Genie 2 Mixer and the Vortex-Genie Pulse |
---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15507355
|
Scientific Industries SI™
SI-H506 |
50 mL Tubes |
105.00 EUR
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15517355
|
Scientific Industries SI™
SI-H512 |
12 x 15 mL Tubes |
119.00 EUR
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15527355
|
Scientific Industries SI™
SI-H524 |
24 Microtubes |
136.00 EUR
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10286369
|
Scientific Industries SI™
SI-H506 |
50 mL Tubes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10296369
|
Scientific Industries SI™
SI-H524 |
24 Microtubes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Holds tubes in place horizontally.
- Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
- No additional attachments are necessary
- Holds tubes in place horizontally.
- Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
- No additional attachments are necessary
Specifications
Vortex-Genie 2 Mixer and the Vortex-Genie Pulse |