Learn More
Scientific Industries™ Horizontal Plastic Clip Microtube Holder
Product Code. 15907965
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15907965 Vortex-Genie Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15907965 Lieferant Scientific Industries™ Lieferanten-Nr. 83041273
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24 microtube holder for Vortex-Genie.
Tube Holder holds twenty-four microtubes horizontally. Holder snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Spezifikation
Model SI-HP524
For Use With (Equipment) Vortex-Genie