Learn More
Honeywell™ Plug ESD S1P Shoe
Specifications
Specifications
| Certifications/Compliance | ISO 9001/2000, Class 2, EC Certified, EN ISO 20345: 2011 S1P SRC, EN 61340-4-3:2001 |
| Closure Type | Velcro strips |
| Footbed Material | Dual Density Polyurethane |
| For Use With (Application) | For microelectronics, semiconductors and printed circuit areas |
| Material | Leather |
| Size | 45 |
| Sole Features | Ensures easy work and flexibility, absorbs shock and protects against temperature changes |
| Sole Material | Dual Density Polyurethane |
| Style | Plug |
| Resistant To | Acids and alkalis, water, abrasion, slip |
| Show More |