Learn More
Sperian™ Honeywell™ Premium Disposable Respirators
Click to view available options
Filter Type:
FFP1
FFP2
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 20
Description
- Rigid shell so shape is maintained even in moist and heavy loading environments
- Hyopallergenic flange seal with absorption properties
- Preformed and colored nose bridge to save time
- Large internal volume to easily communicate and breathe
- Twin elastic straps, ultra flexible and resistant, stapled outside of the filtering area
- No latex, no silicone, no PVC
- Protection and wearer acceptance guaranteed during periods of extended use
Specifications
Specifications
| Certifications/Compliance | EN 149:2001A1:2009, ISO 9001 / 2000, EC |
| Disposable | Single-use |
| Filter Type | FFP1 |
| Product Type | Respiratory Protection |
| Size | Medium |
| With Exhalation Valve | No |
| For Use With (Application) | Textile, construction, iron and steel industries, etc |
| Latex Free | Yes |
| Model | 5110 M/L Respirator |
| Quantity | 20/Pk. |
| Show More |