missing translation for 'onlineSavingsMsg'
Learn More

Homatropine methylbromide, MedChemExpress

Product Code. 16478746
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16478746

Brand: Medchem Express HYB1388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Homatropine methylbromide (Homatropine methobromide) is muscarinic AChR antagonist, inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162.5 nM and 170.3 nM, respectively.

Homatropine methylbromide (Homatropine methobromide) is muscarinic AChR antagonist, inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162.5 nM and 170.3 nM, respectively.

Specifications

Chemical Name or Material Homatropine methylbromide
CAS 80-49-9
Color White
Purity Grade Notes Research
Molecular Formula C17H24BrNO3
Quantity 100 mg
Synonym Homatropine methobromide
Solubility Information DMSO : ≥ 31 mg/mL (83.72 mM)
SMILES C[N+]1([C@@H]2C[C@@H](OC(C(O)C3=CC=CC=C3)=O)C[C@H]1CC2)C.[Br-]
Molecular Weight (g/mol) 370.28
Formula Weight 370.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.