missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Product Code. p-7048700
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346358 25 g 25g
11350999 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346358 Supplier Thermo Scientific Alfa Aesar Supplier No. 011277.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14914-84-2 | Cl3H12HoO6 | 379.37 g/mol

Holmium(III) chloride hexahydrate is used as laboratory reagent and also used in proteomics research , optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14914-84-2
Molecular Formula Cl3H12HoO6
Molecular Weight (g/mol) 379.37
MDL Number MFCD00149739
InChI Key BUQFCXABQYNXLP-UHFFFAOYSA-K
PubChem CID 16211475
SMILES O.O.O.O.O.O.Cl[Ho](Cl)Cl

Specifications

Melting Point 718°C
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula HoCl3·6H2O
Merck Index 14,4727
Solubility Information Soluble in water.
IUPAC Name trichloroholmium;hexahydrate
Formula Weight 379.38 (271.29 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) chloride hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Holmium(III) chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Holmium(III) chloride hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.