Learn More
Kinesis Hollow cathode lamp chromium varian coded 37mm
Brand:  Kinesis 11949-84
Hollow cathode lamp chromium varian coded 37mm
Product Code. 13003570
  • ONLINE EXCLUSIVE PRICE
    531.06 EUR
    476.28 EUR / Each
    Save 54.78 EUR
    10% Off