Learn More
Hoefer™ Wonder Wedge Plate Separation Tool
Product Code. 10390445 Shop All Cytiva Products
Change view
Click to view available options
Length (Metric):
15 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10390445 15 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10390445 Supplier Hoefer™ Supplier No. SE1514
Only null left
Add to basket
Add to basket
Graded notches along one side can be used to determine spacer thickness
  • Plastic, ruler-shaped
  • 15 cm long with tapered ends
  • Notches used to determine spacer thickness, from 0.25 to 3.0 mm
TRUSTED_SUSTAINABILITY
Specifications
Type Plate Separation Tool
Material Plastic
Length (Metric) 15 cm