Includes
- Models SG 15, SG 30, SG 50, and SG 100 include gradient maker with PTFE valves, Luer outlet fitting, support rod, adapter barbed fitting, 22-gauge needle.
- Model SG 500 includes gradient maker with push-pull valve, four wrap-and-seal hose clamps.
Models SG 15, SG 30, SG 50, and SG 100
- Milled of heavy acrylic plastic with high-quality, leak-free PTFE-resin valves
- Offer a flat base for stability on magnetic stirrers; support rod can be attached to ring stand for additional stability
- Accept Luer or 4 mm ID tubing; with included adapters can also accept 2 mm ID or 22-gauge tubing
- SG 100 is ideal for the Hoefer Mighty Small™ Gel Casters SE 275 (for four gels) or SE 215 (for 10 gels)
- Made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base
- Uses a simple push-pull valve to open and close the passage between the reservoir and mixing chambers
- Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns
- Ideal for use with the Hoefer SE 615 or SE 915 Multiple Gel Casters
Specifications
Specifications
| For Use With (Equipment) | SE 615, SE 915 Multiple Gel Casters |
| Type | Gradient Maker |
| Includes | Gradient maker w/Push-Pull Valve, four wrap-and-seal hose clamps |
| Volume (English) | 16.9 oz. |
| Volume (Metric) | 500 mL |