Learn More
Hoefer™ SE660 Tall Standard Dual Cooled Vertical Unit
Product Code. 12845836
Click to view available options
Unit Size:
Each
Product Code. 12845836
Brand: Hoefer™ SE660151.5
Only null left
Add to basket
Add to basket
Taller vertical format accommodates 24 cm gels for increased resolution
  • Dual-Cooled
Specifications
Certifications/Compliance EN61010-1, UL61010-1, CSA C22.2 1010.1, CE
Humidity Range Up to 80%
Temperature (Metric) Maximum 45°C
Depth (Metric) 14 cm