Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
Description
- Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
Specifications
| For Use With (Equipment) | MiniVE Vertical Electrophoresis System |
| Type | Lower Buffer Chamber |