Learn More
Hoefer™ Gel Casters for SE 600 and SE 400 Series Vertical Electrophroesis Systems
Product Code. 12044877
Click to view available options
For Use With (Equipment):
SE 600, SE 400 Series Vertical Electrophoresis System
Unit Size:
Each
Product Code. 12044877
Brand: Hoefer™ SE6015
Only null left
Add to basket
Add to basket
Cast multiple gels
  • Easy-to-handle
  • Leak-free
  • Suitable for one or two gels
Specifications
For Use With (Equipment) SE 600, SE 400 Series Vertical Electrophoresis System
Type Dual Gel Caster
Includes Caster stand with 2 cradles and leveling feet, laminated gaskets
No. of Gels 2
Width (English) Top Plate 7.08 in.
Width (Metric) Top Plate 18 cm