Learn More
MedChemExpress Hoechst 33258 analog 6
Specifications
Specifications
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 25 mg/mL (46.58 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 536.71 g/mol |
| CAS | 129244-66-2 |