Learn More
MedChemExpress Hoechst 33258 analog
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Specifications
Specifications
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : ≥ 51 mg/mL (99.88 mM) |
| Molecular Weight (g/mol) | 510.59 g/mol |
| CAS | 258843-62-8 |