missing translation for 'onlineSavingsMsg'
Learn More

Hobit (ZNF683) Mouse anti-Human, Alexa Fluor 647, Clone: Sanquin-Hobit/1, BD

Product Code. 15834879
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15834879 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15834879 Supplier BD Biosciences Supplier No. 566250

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Sanquin-Hobit/1 monoclonal antibody specifically binds to Homolog of Blimp-1 in T cells (Hobit), which is also known as Zinc finger protein 683 (ZNF683). Hobit is a conserved Zn-finger transcription factor that is homologous to Blimp-1. It is expressed in cytotoxic effector-type CD8+ T cells and in some effector memory CD8+ T cells, but is not significantly expressed in naοve or most central memory CD8+ T cells. Hobit is also expressed in natural killer (NK) cells, and NKT cells, but is not expressed in CD4+ T cells, B cells, monocytes, dendritic cells, or neutrophils. Hobit expression is strongly correlated with the expression T-bet and Interferon-γ(IFN-γ) mRNA. Hobit appears to play a role in the differentiation and effector functions of NK cells and effector CD8+ T cells such as in the production of IFN-γ.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hobit (ZNF683)
Classification Monoclonal
Clone Sanquin-Hobit/1
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Gene Alias HOBIT, Homolog of Blimp-1 in T cells, Zinc finger protein 683, ZNF683
Host Species Mouse
Immunogen Human Hobit Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.