missing translation for 'onlineSavingsMsg'
Learn More

HMN-214, MedChemExpress

Product Code. p-200099714
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477687

Brand: Medchem Express HY12045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HMN-214, an orally bioavailable prodrug of HMN-176, is an inhibitor of polo-like kinase-1 (plk1), with antitumor activity.

HMN-214, an orally bioavailable prodrug of HMN-176, is an inhibitor of polo-like kinase-1 (plk1), with antitumor activity.

Specifications

Chemical Name or Material HMN-214
CAS 173529-46-9
Color White
Purity Grade Notes Research
Molecular Formula C22H20N2O5S
Quantity 5 mg
Synonym IVX-214
Solubility Information DMSO : 25 mg/mL (58.90 mM; Need ultrasonic)
SMILES CC(N(C1=C(C=CC=C1)/C=C/C2=CC=N(C=C2)=O)S(C3=CC=C(C=C3)OC)(=O)=O)=O
Molecular Weight (g/mol) 424.47
Formula Weight 424.47
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.