missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 EUR - 550.00 EUR
Specifications
| Antigen | HMGB2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229691
|
Novus Biologicals
NBP3-35743-100ul |
100 μL |
550.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227391
|
Novus Biologicals
NBP3-35743-20ul |
20 μL |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGB2 Polyclonal antibody specifically detects HMGB2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| HMGB2 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 3148 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| High mobility group protein 2, high mobility group protein B2, high-mobility group box 2, HMG-2, HMG2high-mobility group (nonhistone chromosomal) protein 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human HMGB2 (NP_002120.1).,, Sequence:, SAFFLFCSEHRPKIKSEHPGLSIGDTAKKLGEMWSEQSAKDKQPYEQKAAKLKEKYEKDIAAYRAKGKSEAGKKGPGRPTGSKKKNEPEDEEEEEEEEDED | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title