missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Mouse anti-Human, Alexa Fluor 750, Clone: 115603, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1690S-100UG
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| HMGB1/HMG-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P09429 | |
| HMGB1 | |
| E. coli-derived recombinant human HMGB1/HMG-1 Gly2-Glu215 (predicted) Accession # P09429 | |
| 100 μg | |
| Primary | |
| Detects human HMGB1/HMG-1 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 115603 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3146 | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)