missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA ABC Antibody (Bra-23/9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20835JF646
This item is not returnable.
View return policy
Description
HLA ABC Monoclonal antibody specifically detects HLA ABC in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| HLA ABC | |
| Monoclonal | |
| Janelia Fluor 646 | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Human REH cells (pre-B leukaemia cell line) | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence | |
| Bra-23/9 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction