missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 Protease Monoclonal Antibody (1696)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119015
This item is not returnable.
View return policy
Description
This antibody recognizes free N-terminus of mature HIV protease (HIV-1 and HIV-2), an enzyme that hydrolyzes polyproteins of HIV viruses into functional proteins. The antibody 1696 does not react with the precursor. Functional Application: Inhibition.
A protease is an enzyme that cleaves proteins to their component peptides. The HIV protease (PR) hydrolyzes viral polyproteins into functional protein products that are essential for viral assembly and subsequent activity. This maturation process occurs as the virion buds from the host cell. HIV protease inhibitors are used in the treatment of patients with AIDS and were considered the first breakthrough in over a decade of AIDS research. HIV protease inhibitors can lower the viral load carried by AIDS patents.
Specifications
| HIV1 Protease | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Functional Assay, Western Blot | |
| 1696 | |
| Unconjugated | |
| HIV protease; HIV1gp1 | |
| Bacterially expressed full-length HIV-1 protease. | |
| 100 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction