missing translation for 'onlineSavingsMsg'
Learn More

Histrelin Acetic Acid Salt, TRC

Product Code. 30136162
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136162 2.5 mg 2.5mg
30132394 25 mg 25mg
30007157 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136162 Supplier Toronto Research Chemicals Supplier No. TRCH4563102.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C66H86N18O12 • xCH3COOH
Molecular Weight (g/mol) 1323.506005
Synonym 6-[1-(Phenylmethyl)-D-histidine]-9-(N-ethyl-L-prolinamide)-1-9-luteinizing hormone-releasing factor (Swine);
SMILES O=C(N[C@H](C(N[C@@H](CC1=CC=C(O)C=C1)C(N[C@@H](C(N[C@@H](CC(C)C)C(N[C@@H](CCCNC(N)=N)C(N2CCCC2C(NCC)=O)=O)=O)=O)CC3=CN(CC4=CC=CC=C4)C=N3)=O)=O)CO)[C@@H](NC([C@H](CC5=CN=CN5)NC(C6CCC(N6)=O)=O)=O)CC7=CNC8=CC=CC=C87

Specifications

Analyte or Component Names Histrelin Acetic Acid Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C66H86N18O12/c1-4-70-64(95)55-17-11-25-84(55)65(96)48(16-10-24-71-66(67)68)76-58(89)49(26-38(2)3)77-62(93)53(30-43-34-83(37-74-43)33-40-12-6-5-7-13-40)81-59(90)50(27-39-18-20-44(86)21-19-39)78-63(94)54(35-85)82-60(91)51(28-41-31-72-46-15-9-8-14-45(41)46)79-61(92)52(29-42-32-69-36-73-42)80-57(88)47-22-23-56(87)75-47/h5-9,12-15,18-21,31-32,34,36-38,47-55,72,85-86H,4,10-11,16-17,22-30,33,35H2,1-3H3,(H,69,73)(H,70,95)(H,75,87)(H,76,89)(H,77,93)(H,78,94)(H,79,92)(H,80,88)(H,81,90)(H,82,91)(H4,67,68,71)/t47?,48-,49-,50-,51-,52-,53+,54-,55?/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 2.5 mg
Chemical Name or Material Histrelin Acetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.